Quick connect torch modules for PerkinElmer
Quick-change torch module. Includes inert Scott spray chamber and GemTip Cross-Flow II nebuliser, for Optima Optima 2×00/4×00/5×00/7×00DV
Katalognr. HEWL8003-0340
Anbieter: AGILENT HEWLETT PACKARD
Quick connect torch modules for PerkinElmer
Katalognr. HEWL8003-0340
Anbieter: AGILENT HEWLETT PACKARD
Direkt zu
Product Details & Documents
The Quick-Connect torch module for PerkinElmer ICP-OES includes the torch, mounting block, 2 mm alumina injector, Scott spray chamber, and Cross-Flow nebuliser.
- Guaranteed to be compatible with PerkinElmer instruments
- Backed by the stand behind warranty that guarantees the parts will not cause instrument failure or downtime
- Parts qualified on a genuine PerkinElmer system
- Many parts in packaging designed to be more durable, sturdy and provide the best protection during shipping to ensure the part arrives with no damage
The Quick-Change module is useful when you are running different sample types and need separate sample introduction for each.
Technische Daten
- Bezeichnung:Quick-change torch module. Includes inert Scott spray chamber and GemTip Cross-Flow II nebuliser, for Optima Optima 2×00/4×00/5×00/7×00DV
- Seq:06