Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Quick connect torch modules for PerkinElmer
Quick connect torch modules for PerkinElmer
Mounting block, for quick-change torch, does not include torch, injector, spray chamber, or nebuliser, for Optima Optima 2×00/4×00/5×00/7×00/8×00
Catalog # HEWL8003-0338
Quick connect torch modules for PerkinElmer
Catalog # HEWL8003-0338
Quick connect torch modules for PerkinElmer
Mounting block, for quick-change torch, does not include torch, injector, spray chamber, or nebuliser, for Optima Optima 2×00/4×00/5×00/7×00/8×00
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
The Quick-Connect torch module for PerkinElmer ICP-OES includes the torch, mounting block, 2 mm alumina injector, Scott spray chamber, and Cross-Flow nebuliser.
  • Guaranteed to be compatible with PerkinElmer instruments
  • Backed by the stand behind warranty that guarantees the parts will not cause instrument failure or downtime
  • Parts qualified on a genuine PerkinElmer system
  • Many parts in packaging designed to be more durable, sturdy and provide the best protection during shipping to ensure the part arrives with no damage
The Quick-Change module is useful when you are running different sample types and need separate sample introduction for each.
Specifications
  • Bezeichnung:
    Mounting block, for quick-change torch, does not include torch, injector, spray chamber, or nebuliser, for Optima Optima 2×00/4×00/5×00/7×00/8×00
  • Seq:
    01
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.