ContactUsLinkComponent
SAF SAFE SEAL SURPHOB LACE, 300 UL, STERILE, FILTER TIPS LOW BINDING, GRADUATED, DNA, DNASE, RNASE, PYROGENIC FREE 1 * 960 ST
Katalognr. BZYMVT0250
Anbieter:  BIOZYM
SAF SAFE SEAL SURPHOB LACE, 300 UL, STERILE, FILTER TIPS LOW BINDING, GRADUATED, DNA, DNASE, RNASE, PYROGENIC FREE 1 * 960 ST
Katalognr. BZYMVT0250
Anbieter:  BIOZYM
Frequently Bought Together