Pierce™ Protein Concentrators
Product Details & Documents
Protein Concentrators PES are disposable ultrafiltration centrifugal devices with a polyethersulfone (PES) membrane for concentration, desalting, and buffer exchange of biological samples such as proteins and nucleic acids.
- Sample concentration of 10- to 30-fold can be achieved in 10 minutes or less
- Unique design minimises membrane fouling and provides high speed concentrations, even with particle laden solutions
- Compatible with most bench-top microcentrifuges with fixed-angle rotors that accommodate 2,2 ml tubes
- Molecular weight cutoff
- Can be used for processing sample volumes
- Rapid protein concentration
- Excellent protein recovery
- Reliable, consistent results
The 0,5 ml concentrators contain a vertical PES membrane, which allows for processing of small volumes between 100 and 500 µl. The MWCOs are etched on the sides of the concentrators for easy identification, while a clear window with graduations on the side of each device allows for visual determination of the retentate volume. Typical protein recovery for proteins that are ≥ two-fold greater than the MWCO is >90%, the minimum protein sample concentration should be 0,1 mg/ml.
The 2 to 6 ml concentrators have a built-in dead stop and contain a vertical low protein-binding, high flux PES membrane, which allows for processing of volumes between 2 and 6 ml. The MWCOs are etched on the sides of the concentrators for easy identification, while a clear window with graduations on the side of each device allows for visual determination of the retentate volume.
Pierce™ Protein Concentrators PES contain a membrane rated for retaining molecules with a molecular weight at least twofold greater than the MWCO rating of the PES membrane within the device. Reduced recovery may occur when using a concentrator with molecules smaller than the recommended mass. Protein recovery will vary depending on the specific protein in the sample and its starting concentration.
Caution: For research use only. Not for use in diagnostic procedures.
Technische Daten
- MWCO:30 kD
- Bezeichnung:Pierce™ Protein concentrator PES, 20 - 100 ml
- Seq:23
- Cat. No.:PIER88536
- Temperatur:RT
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



