Multi-gas detector, GasAlert Micro 5
O₂, H₂S, LEL, 0 - 30 % (O₂) 0 - 500 ppm (H₂S) 0 - 100 % LEL
Katalognr. BWTEM5XWH0RDDYU00
Direkt zu
Artikel gestrichen
Product Details & Documents
GasAlert Micro 5 was designed for protecting five potential atmospheric hazards including oxygen, combustible and toxic gases.
- Integral concussion-proof boot
- Four alarm levels: Low, High, TWA and STEL
- Compatible with MicroDock II automatic test and calibration station
It continuously monitors and displays concentrations of up to five gases.
Lieferinformationen: Supplied with a detector complete with specified sensor(s), stainless steel alligator clip, concussion-proof housing, cradle charger, wall outlet charging adaptor, screwdriver, calibration/test adaptor, hose, manual and multi-language CD manual.
Technische Daten
- Parameter:O₂, H₂S, LEL
- Messbereich:0 - 30 % (O₂) 0 - 500 ppm (H₂S) 0 - 100 % LEL
- Seq:08
- Cat. No.:BWTEM5XWH0RDDYU00
- Modell:Micro 5
- B×T×H:145×74×38 mm
- Akku-Typ:Rechargeable
- Alarm:Visual, vibrating, audible (95 dB)
- Batterielebensdauer:20 hours
- Gewicht:370 g
- Temperaturbereich:−20…+50 °C
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



