Multi-gas detector, GasAlert Micro 5
O₂, H₂S, CO, LEL, 0 - 30 % (O₂) 0 - 500 ppm (H₂S) 0 - 999 ppm (CO) 0 - 100 % LEL
Katalognr. BWTEM5XW0YRDDYU00
Direkt zu
Artikel gestrichen
Product Details & Documents
GasAlert Micro 5 was designed for protecting five potential atmospheric hazards including oxygen, combustible and toxic gases.
- Integral concussion-proof boot
- Four alarm levels: Low, High, TWA and STEL
- Compatible with MicroDock II automatic test and calibration station
It continuously monitors and displays concentrations of up to five gases.
: Supplied with a detector complete with specified sensor(s), stainless steel alligator clip, concussion-proof housing, cradle charger, wall outlet charging adaptor, screwdriver, calibration/test adaptor, hose, manual and multi-language CD manual.
Technische Daten
- Parameter:O₂, H₂S, CO, LEL
- Messbereich:0 - 30 % (O₂) 0 - 500 ppm (H₂S) 0 - 999 ppm (CO) 0 - 100 % LEL
- Seq:06
- Cat. No.:BWTEM5XW0YRDDYU00
- Modell:Micro 5
- B×T×H:145×74×38 mm
- Akku-Typ:Rechargeable
- Alarm:Visual, vibrating, audible (95 dB)
- Batterielebensdauer:20 hours
- Gewicht:370 g
- Temperaturbereich:−20…+50 °C



