Dichtungen
Piston seal wash seal/priming valve seal, ICS-3000, ICS-5000, 063382, DION
CTSA11-2587
: SCIENCIX
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Sciencix parts are equivalent to the corresponding OEM part.
- Bezeichnung:Piston seal wash seal/priming valve seal
- Seq:23
- Erstausrüster:DION
- Erstausrüster-Äquivalent:063382
- Modell:ICS-3000, ICS-5000