AuftragsverfolgungKontakt
Microplates, micro-volume, SpectraDrop™
Microplates, micro-volume, SpectraDrop™
SpectraDrop™ 64-Well-Nachfüllkit für Niedrigvolumen-Objektträger
N° de catalogue MLDV0200-6264
Fournisseur:   Molecular Devices
Microplates, micro-volume, SpectraDrop™
N° de catalogue MLDV0200-6264
Fournisseur:  Molecular Devices
Microplates, micro-volume, SpectraDrop™
Microplates, micro-volume, SpectraDrop™
SpectraDrop™ 64-Well-Nachfüllkit für Niedrigvolumen-Objektträger
Prix
SFr. 1’584.00
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
SpectraDrop™ micro-volume microplate consists of aspecially designed adapter, PTFE-coated bottom slide, and top slide with evaporation-reducing spacers.
  • Handle volumes down to 2 µl
  • Used to quantify DNA, RNA and protein
  • Cleaning convenience with hasslefree handling
  • PTFE-coated to ensure precise droplet placement
  • Slide design is compatible with 8- and/or 16-channel pipettors
These microplates comes in two density configurations, 24-wells per plate and 64-wells per plate, with two volume options, 2 or 4 µl, allowing users to select the configuration ideal for their needs.
Informations de livraison: The kit includes one microplate adapter, two 24-well low volume bottom slides, and one of each cover slide, 2 and 4 µl (SpectraDrop™ 24 kit) and one microplate adapter, five 24-well low volume slides, five 64-well low volume bottom slides and five of each cover slide, 2 and 4 µl (SpectraDrop™ HTS kit).
Spécifications
  • Bezeichnung:
    SpectraDrop™ 64-Well-Nachfüllkit für Niedrigvolumen-Objektträger
  • Seq:
    04