Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Microplates, micro-volume, SpectraDrop™ | Avantor
Microplates, micro-volume, SpectraDrop™
SpectraDrop™ 24-Well-Nachfüllkit für Niedrigvolumen-Objektträger
Katalognr. MLDV0200-6263
Anbieter:   Molecular Devices
Microplates, micro-volume, SpectraDrop™
Katalognr. MLDV0200-6263
Anbieter:  Molecular Devices
Microplates, micro-volume, SpectraDrop™
Microplates, micro-volume, SpectraDrop™
SpectraDrop™ 24-Well-Nachfüllkit für Niedrigvolumen-Objektträger
Preis
SFr. 643.00
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
SpectraDrop™ micro-volume microplate consists of aspecially designed adapter, PTFE-coated bottom slide, and top slide with evaporation-reducing spacers.
  • Handle volumes down to 2 µl
  • Used to quantify DNA, RNA and protein
  • Cleaning convenience with hasslefree handling
  • PTFE-coated to ensure precise droplet placement
  • Slide design is compatible with 8- and/or 16-channel pipettors
These microplates comes in two density configurations, 24-wells per plate and 64-wells per plate, with two volume options, 2 or 4 µl, allowing users to select the configuration ideal for their needs.
Lieferinformationen: The kit includes one microplate adapter, two 24-well low volume bottom slides, and one of each cover slide, 2 and 4 µl (SpectraDrop™ 24 kit) and one microplate adapter, five 24-well low volume slides, five 64-well low volume bottom slides and five of each cover slide, 2 and 4 µl (SpectraDrop™ HTS kit).
Technische Daten
  • Bezeichnung:
    SpectraDrop™ 24-Well-Nachfüllkit für Niedrigvolumen-Objektträger
  • Seq:
    03
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.