Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Stirring paddles | Avantor

Stirring paddles

Stirring paddle, with solid blade, 1500 mm, 300×150 mm
Katalognr. SAMPA344-1500
Anbieter:   Sampling Systems
Stirring paddles
Katalognr. SAMPA344-1500
Anbieter:  Sampling Systems
Stirring paddles
Stirring paddles

Stirring paddle, with solid blade, 1500 mm, 300×150 mm

Preis
SFr. 236.00
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents

These paddles are ideal for use when producing liquids and suspensions. Stirrers are made from 316 stainless steel

  • Robust construction
  • Crevice free
  • Satin finish
  • BSE/TSE free
Technische Daten
  • Bezeichnung:
    Stirring paddle, with solid blade
  • Länge:
    1500 mm
  • Seq:
    02
  • Klinge L×B:
    300×150 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.