Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Spore ampoules | Avantor

Spore ampoules

Geobacillus stearothermophilus cell line 7953: negative control ampoule (24 month shelf life) / sterilisation process: steam liquid cycles, 58 mm, 1 ml
Katalognr. NAMSSA1-NC-10
Anbieter:   CROSSTEX
Spore ampoules
Katalognr. NAMSSA1-NC-10
Anbieter:  CROSSTEX
Spore ampoules

Geobacillus stearothermophilus cell line 7953: negative control ampoule (24 month shelf life) / sterilisation process: steam liquid cycles, 58 mm, 1 ml

Preis
SFr. 65.40
Einloggen für Verfügbarkeit
1 * 1 Un.
Product Details & Documents

These spore ampoules are ideal for use in validation and routine monitoring of liquid steam sterilization cycles. Spore ampoules are manufactured using hermetically sealed Type I borosilicate glass containing an exclusively formulated Tryptic
Soy Broth (TSB) modified with pH indicator, and Geobacillus stearothermophilus spores. No activation is required; simply expose Spore Ampoules to sterilization process and incubate.

  • Reduced incubation time of 48 hours
  • Clear visible results as growth is evident by turbidity and/or a color shift of the media from purple to yellow
  • Saves time by eliminating aseptic culturing required with traditional spore strips
  • Unique packaging insert keeps ampoules in upright position and allows for quick reference to the number of units remaining

 : Each shipment is accompanied by a certificate of analysis and instructions for use

 : Storage under refrigerated conditions (2 to 8 °C) is required. Do not freeze spore ampoules.

Dokumente

Technische Daten
  • Volumen:
    1 ml
  • Bezeichnung:
    Geobacillus stearothermophilus cell line 7953: negative control ampoule (24 month shelf life) / sterilisation process: steam liquid cycles
  • Länge:
    58 mm
  • Seq:
    07
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.