Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
pH/mV/°C/ISE meter, handheld, Eutech pH 450

pH/mV/°C/ISE meter, handheld, Eutech pH 450

pH/mV/ISE meter, handheld, Eutech pH 450 (meter only)
Catalog # 662-2301
Supplier:   EUTECH
pH/mV/°C/ISE meter, handheld, Eutech pH 450
Catalog # 662-2301
Supplier:  EUTECH
pH/mV/°C/ISE meter, handheld, Eutech pH 450
pH/mV/°C/ISE meter, handheld, Eutech pH 450

pH/mV/ISE meter, handheld, Eutech pH 450 (meter only)

Price
555.00 SFr.
Log in for availability
1 * 1 ST
Product Details & Documents

This portable meter is ideal for use in any environment. It features a durable waterproof and dustproof design with IP67 rating.

  • Large, backlit display with intuitive user-friendly icons
  • Grip-Clip™: Quickly and easily secures the pH electrode to a beaker or container
  • Automatic buffer recognition; up to 5- or 6-point calibration; calibration due alarm
  • Stores up to 500 data sets; easy USB or RS232 output with data cables (cables not always included)
  • Meets GLP standards: Built-in real-time clock stamps stored data and calibration data with time and date
  • Can be used with two AA batteries or with optional universal power adapter (not included)
  • Built-in stand with wall-mount option allows for easy bench top measurement or storage

The meter can be used with pH, ORP or ISE electrodes with BNC connector.

Delivery information: Meter only (without electrodes) is supplied with Grip-Clip™ and two AA batteries.
Set 1 additionally includes a double junction pH electrode and ATC probe.
Set 2 additionally includes a double-junction pH electrode and ATC probe, hard carry case, USB cable, pH buffers, storage solution and rinse solution.
Set 3 additionally includes a double-junction pH/ATC electrode, hard carry case, USB cable, pH buffers, storage solution and rinse solution.

Documents

Specifications
  • Bezeichnung:
    pH/mV/ISE meter, handheld, Eutech pH 450 (meter only)
  • Seq:
    01
  • ISE Kalibrierung:
    From 2 - 4 or 2 - 5 points: 0,10 / 1,00 / 10,0 / 100,0 / 1000
    0,20 / 2,00 / 20,0 / 200,0 / 2000
    0,50 / 5,00 / 50,0 / 500,0
  • Temperatur - Kompensation:
    Automatic or manual from −10 to +110 °C
  • mV - Bereich:
    ±2000
  • mV - Genauigkeit:
    ±0,2 +2 LSD or +0,5 % of reading; whichever is greater
  • ISE Genauigkeit:
    ±0,5 % full scale (monovalent); ±1 % full scale (divalent)
  • Ausgänge:
    USB, RS232
  • Temperaturgenauigkeit:
    ±0,5 °C
  • Temperaturauflösung:
    0,1 °C
  • ISE Auflösung:
    0,01 / 0,1 / 1
  • Konformität:
    IP67
  • Temperatur-Kalibrierung:
    Offset in 0,1 increments, ±10 °C
  • Gewicht:
    0,5 kg
  • pH - Genauigkeit:
    ±0,01
  • pH - Bereich:
    −2,00...+16,00
  • mV - Kalibrierung:
    ±200
  • Cat. No.:
    662-2301
  • Modell:
    pH 450
  • Kapazität Datenprotokollierung:
    500 data sets
  • B×T×H:
    200×83×57 mm
  • pH - Auflösung:
    0,01
  • mV - Auflösung:
    0,1 (±999,9); 1 (beyond)
  • Stromversorgung:
    Two AA batteries (included), AC adapter (optional)
  • ISE Bereich:
    0,0 - 2000 ppm
  • pH - Kalibrierung:
    Up to 5 points (USA & NIST & CUSTOM); up to 6 points (DIN)
  • Temperaturbereich:
    −10,0...+110,0 °C
  • Display:
    Custom LCD with backlight
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.