Labels, hazard warning, electricity
Hazard labels, PIC 329-TRI 100-B7541, Polyester, Schwarz auf gelb, 100×87 mm, 3 labels/card
Katalognr. BRDY250288
Anbieter: Brady
Direkt zu
Product Details & Documents
These electricity hazard warning labels are made of polyester and are used for prevention of accidents.
- Self-adhesive
- Clear identification of voltage, phases, grounding and insulation
Dokumente
Technische Daten
- Farbe:Schwarz auf gelb
- Bezeichnung:Hazard labels
- Abmessungen:100×87 mm
- Material:Polyester
- Seq:06
- Modell:PIC 329-TRI 100-B7541
- Verpackt:3 labels/card