ContactUsLinkComponent
For rapid testing for changes in the properties of materials caused by sunlight, temperature and humidity. Product changes that take months or years indoors or outdoors, such as fading, yellowing or embrittlement, can often be simulated in a Suntest 1 * 1 ST
Katalognr. MSPP55008040
For rapid testing for changes in the properties of materials caused by sunlight, temperature and humidity. Product changes that take months or years indoors or outdoors, such as fading, yellowing or embrittlement, can often be simulated in a Suntest 1 * 1 ST
Katalognr. MSPP55008040
Frequently Bought Together