Combustion boats
30×10×5 mm
Katalognr. TSLA75141
Anbieter: TECHNICAL QUARTZ SOLUTIONS
Direkt zu
Product Details & Documents
Combustion boats, quartz, glazed, with or without handle.
- Very good chemical resistance
- Very good temperature resistance
Technische Daten
- Seq:04
- B×T×H:30×10×5 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.