Combustion boats
25×5×4 mm
Katalognr. TSLA75140
Anbieter: TECHNICAL QUARTZ SOLUTIONS
Direkt zu
Product Details & Documents
Combustion boats, quartz, glazed, with or without handle.
- Very good chemical resistance
- Very good temperature resistance
Technische Daten
- Seq:03
- B×T×H:25×5×4 mm