Bench mounted gas taps
2× propane/butane (LPG), EPDM seal
Katalognr. KOTT733-00056
Anbieter: KOETTERMANN
Direkt zu
Product Details & Documents
These bench mounted gas taps are sealed by EPDM (ethylene propylene diene terpolymer) which is an extremely durable synthetic rubber roofing membrane.
Technische Daten
- Bezeichnung:2× propane/butane (LPG), EPDM seal
- Seq:02