AuftragsverfolgungKontakt
Sterilisation Boxes for Pipettes | Avantor
Sterilisation Boxes for Pipettes
Pipette box for the storage of pipettes (315 - 485 mm)
Katalognr. BRND28935
Anbieter:   Brand
Sterilisation Boxes for Pipettes
Katalognr. BRND28935
Anbieter:  Brand
Sterilisation Boxes for Pipettes
Sterilisation Boxes for Pipettes
Pipette box for the storage of pipettes (315 - 485 mm)
Preis
92.30 SFr.
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
These pipette boxes made of aluminum are ideal for the storage and sterilisation of pipettes with different lengths.
  • With inner silicone coating
  • Very good temperature resistance
  • Very good chemical resistance
Technische Daten
  • Bezeichnung:
    Pipette box for the storage of pipettes (315 - 485 mm)
  • Seq:
    04