Software für Spektrometer
Datrys™ CFR Software, Ultrospec™ 7000/8000/9000
Katalognr. SCLI29003624
Anbieter: BIOCHROM
Direkt zu
Product Details & Documents
Die Datrys Software wird in verschiedenen Modulen geliefert, um die Anwendungsanforderungen verschiedener Kundengruppen zu erfüllen, und bietet einfache Erweiterungsmöglichkeiten, falls sich Ihre Anforderungen ändern sollten.
- Datrys (standard) software for routine measurement analysis and PC control
- Datrys Lite software for quick read, fast scanning and PC control
- Datrys Life Science software for nucleic acids, proteins and cell density measurement analysis and PC control
- Datrys CFR software supports 21 cfr part 11 compatibility, routine measurement analysis and PC control
For many users, having an external PC to control their spectrophotometer and manipulate data, gives them the ultimate in flexibility and control. Whether looking for small differences in multiple spectral overlays, carrying out post-run manipulations on large numbers of samples, Datrys software has the flexibility to work in the way you want.
Dokumente
Infotable 9581385 Software for Spectrophotometers Biochrom
Technisches Datenblatt
Technische Daten
- Bezeichnung:Datrys™ CFR Software
- Seq:04
- Für:Ultrospec™ 7000/8000/9000