Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Tank liners, Corning®

Tank liners, Corning®

200 L, LDPE, 55,88×91,44 mm
Catalog # MDTC91-300-35
Supplier:   Corning
Tank liners, Corning®
Catalog # MDTC91-300-35
Supplier:  Corning
Tank liners, Corning®

200 L, LDPE, 55,88×91,44 mm

Price
SFr. 92.40
Log in for availability
1 * 1 ST
Product Details & Documents

Corning® tank liners are designed to fit cylindrical tanks.

  • Available in a range of sizes

Tank liners are manufactured from LDPE.

Specifications
  • Ø×H:
    55,88×91,44 mm
  • Kapazität:
    200 L
  • Seq:
    05
  • Schlauchmaterial:
    LDPE
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.