Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
 

CCVJ ≥95%

10 mg
Katalognr. CAYM17870-10
Anbieter:   Cayman Chemical
CCVJ ≥95%
Katalognr. CAYM17870-10
Anbieter:  Cayman Chemical
CAS Nummer:  142978-18-5
CCVJ ≥95%

10 mg

CAS Nummer:  142978-18-5
Preis
363.00 SFr.
Einloggen für Verfügbarkeit
1 * 10 mg
Product Details & Documents

CCVJ is a fluorescent molecular rotor characterised by low background fluorescence, low fluorescence anisotropy, and good water solubility. Fluorescent molecular rotors are molecules whose fluorescence is inversely proportional to its intramolecular rotation.

The intramolecular rotation of these probes, and hence their fluorescence, depends on the immediate microenvironment of the probe. As a result, fluorescent molecular rotors are used to evaluate changes in solution and membrane viscosity, polymerisation or aggregation processes, and protein (un)folding.

Moreover, it has large hydrophobic structures, allowing it to associate in a non-covalent manner with hydrophobic pockets in proteins in solution. It is broadly used to monitor changes in solution and molecular characteristics.

Synonyme:  9-(2-Carboxy-2-cyanovinyl)julolidine, (E)-2-Cyano-3-(1,2,3,5,6,7-hexahydropyrido[3,2,1-ij]quinolin-9-yl)acrylic acid
  • CAS Nummer:142978-18-5
  • Formel:C₁₆H₁₆N₂O₂
  • MDL-Nummer:MFCD00274408
  • Molekulargewicht:268.31 g/mol
Technische Daten

Ergebnisse des Spezifikationstests

  • Purity:
    ≥95%
  • Formal Name:
    2-cyano-3-(2,3,6,7-tetrahydro-1H,5H-benzo[ij]quinolizin-9-yl)-2-propenoic acid
  • Formulation:
    A Crystalline Solid
  • λmax:
    279, 441 nm
  • SMILES:
    OC(/C(C#N)=C/C1=CC(CCC2)=C(N2CCC3)C3=C1)=O
  • InChI Code:
    1S/C16H16N2O2/c17-10-14(16(19)20)9-11-7-12-3-1-5-18-6-2-4-13(8-11)15(12)18/h7-9H,1-6H2,(H,19,20)/b14-9+
  • InChI Key:
    JXENNHTVELFRHV-NTEUORMPSA-N
  • Storage:
    −20 °C
  • Shipping:
    Room temperature in continental US; may vary elsewhere

Technische Daten

  • VE:
    10 mg

Technische Daten