Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Choline dihydrogen citrate ≥98% | Avantor

Choline dihydrogen citrate ≥98%

Poly pail, 12 kg
Katalognr. SPCMC1229-12KG
Reizend
Anbieter:   Spectrum Chemical
Choline dihydrogen citrate ≥98%
Katalognr. SPCMC1229-12KG
Anbieter:  Spectrum Chemical
CAS Nummer:  77-91-8
Choline dihydrogen citrate ≥98%

Poly pail, 12 kg

CAS Nummer:  77-91-8
Preis
SFr. 2’110.00
Einloggen für Verfügbarkeit
1 * 12 kg
Product Details & Documents

Choline Dihydrogen Citrate is part of a class of quaternary ammonium salts containing the N,N,N-trimethylethanolammonium cation and Dihydrogen Citrate counteranion. Choline is the precursor for the neurotransmitter acetylcholine, which is involved in many functions including memory and muscle control. This item is of known assay and suitable for most chemical uses.

GHS hazard : Exclamation mark

Synonyme:  Choline dihydrogencitrate salt, 2-Hydroxy-N,N,N-trimethylethan-1-aminium 3,4-dicarboxy-3-hydroxybutanoate
  • CAS Nummer:77-91-8
  • Formel:C₁₁H₂₁NO₈
  • MDL-Nummer:MFCD00038734
  • Molekulargewicht:295.29 g/mol
  • Lagertemperatur:Raumtemperatur
Technische Daten

Ergebnisse des Spezifikationstests

  • Assay:
    98% min.
  • Water:
    max.0.25 %
  • Residue On Ignition:
    max. 0.05 %

Technische Daten

  • Packung:
    Poly pail
  • VE:
    12 kg

Technische Daten

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.