1-Octadecanol NF
Poly pail, 2,5 kg
Catalog # SPCMST115-2.5KG
Supplier: Spectrum Chemical
Jump to:
Product Details & Documents
BSE/TSE statement (available upon request)
Allergen statement (available upon request)
Synonyms: Stearyl alcohol, 1-Hydroxyoctadecane, Heptadecyl carbinol, 1-Octadecyl alcohol
- CAS Number:112-92-5
- Formula:H₃C(CH₂)₁₆CH₂OH
- MDL Number:MFCD00002823
- Molecular Weight:270.5 g/mol
- Merck Index:12,08960
- Boiling Pt:336 °C (1013 hPa)
- Melting Pt:56…57 °C
- Density:0,805…0.815 g/cm³ (60 °C)
- Storage Temperature:Raumtemperatur
- EINECS:204-017-6
Specifications
Specification Test Results
- Assay:90% Min.
Specifications
- Packung:Poly pail
- VE:2,5 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



