Sucralose NF
Poly pail, 2,5 kg
SPCMS1416-2.5KGBL
All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).
: Trichlorosucrose, (2R,3R,4R,5R,6R)-2-(((2R,3S,4S,5S)-2,5-Bis(chloromethyl)-3,4-dihydroxytetrahydrofuran-2-yl)oxy)-5-chloro-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4-diol, 1-(1,6-Dichloro-1,6-dideoxy-beta-D-fructofuranosyl)-4-chloro-4-deoxy-alpha-D-galactopyranoside
- :56038-13-2
- :C₁₂H₁₉Cl₃O₈
- :MFCD03648615
- :397.63 g/mol
- :130 °C
- :Raumtemperatur
Documents
SPCMS1416-2.5KGBL_TDS_EN
Product specification
- APPEARANCE:CONFORM
- ASSAY (ANHYDROUS BASIS):98.0 - 102.0 %
- SPECIFIC ROTATION:+84.0° to +87.5°
- WATER DETERMINATION:Max. 2.0 %
- RESIDUE ON IGNITION:Max. 0.7 %
- CADMIUM (Cd):AS REPORTED
- LEAD (Pb):AS REPORTED
- ARSENIC (As):AS REPORTED
- MERCURY (Hg):AS REPORTED
- HYDROLYSIS PRODUCTS:Max. 0.1 %
- METHANOL:Max. 0.1 %
- RELATED COMPOUNDS:Max. 0.5 %
- IDENTIFICATION A (FTIR):SPECTRUM MATCHES REFERENCE
- IDENTIFICATION (B):RETENTION TIME CORRESPONDS TO STANDARD
- IDENTIFICATION (C):Rf VALUE CORRESPONDS TO STANDARD
- RESIDUAL SOLVENTS:AS REPORTED
- Packung:Poly pail
- VE:2,5 kg
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



