Sucralose NF
Poly pail, 10 kg
Katalognr. SPCMS1416-10KGBL
Anbieter: Spectrum Chemical
Direkt zu
Product Details & Documents
All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).
Synonyme: Trichlorosucrose, (2R,3R,4R,5R,6R)-2-(((2R,3S,4S,5S)-2,5-Bis(chloromethyl)-3,4-dihydroxytetrahydrofuran-2-yl)oxy)-5-chloro-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4-diol, 1-(1,6-Dichloro-1,6-dideoxy-beta-D-fructofuranosyl)-4-chloro-4-deoxy-alpha-D-galactopyranoside
- CAS Nummer:56038-13-2
- Formel:C₁₂H₁₉Cl₃O₈
- MDL-Nummer:MFCD03648615
- Molekulargewicht:397.63 g/mol
- Schmelzender Platingehalt:130 °C
- Lagertemperatur:Raumtemperatur
Documents
SPCMS1416-2.5KGBL_TDS_EN
Product specification
Technische Daten
Ergebnisse des Spezifikationstests
- APPEARANCE:CONFORM
- ASSAY (ANHYDROUS BASIS):98.0 - 102.0 %
- SPECIFIC ROTATION:+84.0° to +87.5°
- WATER DETERMINATION:Max. 2.0 %
- RESIDUE ON IGNITION:Max. 0.7 %
- CADMIUM (Cd):AS REPORTED
- LEAD (Pb):AS REPORTED
- ARSENIC (As):AS REPORTED
- MERCURY (Hg):AS REPORTED
- HYDROLYSIS PRODUCTS:Max. 0.1 %
- METHANOL:Max. 0.1 %
- RELATED COMPOUNDS:Max. 0.5 %
- IDENTIFICATION A (FTIR):SPECTRUM MATCHES REFERENCE
- IDENTIFICATION (B):RETENTION TIME CORRESPONDS TO STANDARD
- IDENTIFICATION (C):Rf VALUE CORRESPONDS TO STANDARD
- RESIDUAL SOLVENTS:AS REPORTED
Technische Daten
- Packung:Poly pail
- VE:10 kg
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



