Agar, Pulver NF
Fiber drum, 25 kg
Katalognr. SPCMAG110-25KGBL
Anbieter: Spectrum Chemical
Direkt zu
Product Details & Documents
Form: Powder
Synonyme: Agar Agar
- CAS Nummer:9002-18-0
- MDL-Nummer:MFCD00081288
- Merck-Index:13,00184
- Schmelzender Platingehalt:90 °C
- Lagertemperatur:Raumtemperatur
- EINECS:232-658-1
Technische Daten
Technische Daten
- Packung:Fiber drum
- VE:25 kg
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



