Cyclopentanone ≥99%
20 L
Katalognr. SPCMC1124-20LTBL
Anbieter: Spectrum Chemical
Direkt zu
Product Details & Documents
Cyclopentanone is a liquid that has an odor similar to peppermint and is devoid of color. It's an organic compound and a cyclic ketone that most often used as a fragrance.
GHS hazard : Flame;Exclamation mark
UN2245
- CAS Nummer:120-92-3
- Formel:C₅H₈O
- MDL-Nummer:MFCD00001409
- Molekulargewicht:84.12 g/mol
- Dichte:0,951
- Lagertemperatur:Raumtemperatur
- UN:2245
- ADR:3,III
Technische Daten
Ergebnisse des Spezifikationstests
- Assay:99% min.
Technische Daten
- VE:20 L



