AuftragsverfolgungKontakt
Flow-through vessel | Avantor
Flow-through vessel
Flow vessel for field measurements, D 3Sen
Katalognr. WTWA903842
Anbieter:   WTW
Flow-through vessel
Katalognr. WTWA903842
Anbieter:  WTW
Flow-through vessel
Flow-through vessel
Flow vessel for field measurements, D 3Sen
Preis
SFr. 1’090.00
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
Flow vessels for WTW conductivity measuring cells are perfectly designed to perform measurements without the interference of carbon dioxide through the air or to examine the conductivity of solutions in closed circuits.
D 3Sen: Designed for everybody who must perform flow-through measurements in the field, i.e. ground water pumping tests. It consists of a rugged flow-through vessel in holder and a tripod, also suitable for pole mounting. With hose fittings for standard hoses with a diameter of 19 mm. Three places are available for pH/ORP, conductivity and dissolved oxygen sensors (CellOx/FDO). Including mounting accessories.
Achtung! Bitte beachten: Please note: The overpressure in the system must not exceed 0,5 bar.
Dokumente
Technische Daten
  • Bezeichnung:
    Flow vessel for field measurements, D 3Sen
  • Seq:
    04