Lamivudine, Reference Standards
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Lamivudine Resolution Mixture B: This is a mixture of lamivudine (about 94%) and lamivudine diastereomer (about 6%)
: US Pharmacopea Reference Standard
Specifications
Specifications
- Packung:Vial
- VE:0,8 mg
- Normen:USP reference standard
- Bezeichnung:Lamivudine Resolution Mixture C
Specifications
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



