ContactUsLinkComponent
STABILIZER FOR CONVENTIONAL, SULFONATE-BASED METALLO-ORGANIC STANDARDS. ADD TO SOLUTIONS AT 0.6WT%. 1 * 50 g
Katalognr. PROOVHG-STAB-50G
STABILIZER FOR CONVENTIONAL, SULFONATE-BASED METALLO-ORGANIC STANDARDS. ADD TO SOLUTIONS AT 0.6WT%. 1 * 50 g
Katalognr. PROOVHG-STAB-50G
Frequently Bought Together