ContactUsLinkComponent
CHROMIUM 50 IN 2% HNO3, CR: 10 µG/ML NOM., 50CR APPROX. 97% 1 * 50 mL
Katalognr. PROOVHG-LIS50CR-50
CHROMIUM 50 IN 2% HNO3, CR: 10 µG/ML NOM., 50CR APPROX. 97% 1 * 50 mL
Katalognr. PROOVHG-LIS50CR-50
Frequently Bought Together