SULFUR - S @ 100UG/G IN HEAVY MINERAL OIL, 100ML 1 * 100 mL
Katalognr. PROOVHGSMIN100100
Anbieter: LGC Standards PROMOCHEM
SULFUR - S @ 100UG/G IN HEAVY MINERAL OIL, 100ML 1 * 100 mL
Katalognr. PROOVHGSMIN100100
Anbieter: LGC Standards PROMOCHEM
Frequently Bought Together
Desosaminylazithromycin, Reference Standard
Anbieter: USP
Zidovudine, Reference Standards
Anbieter: USP
[EN]SULFAMETHOXAZOLE-D4 1 * 5 mg
Anbieter: LGC Standards PROMOCHEM
CEFAZOLIN SODIUM SALT 1 * 75 mg
Anbieter: Merck
Glyceryl Tricaprylate, Reference Standard
Anbieter: USP
Organic reference standards, Chloroparaffin C10-C13, Dr. Ehrenstorfer
Anbieter: EHRENSTORFER
PYMETROZINE CERTIFIED 1 * 0,1 g
Anbieter: EHRENSTORFER
[EN]DIBROMOCHLOROMETHANE 1 * 1 mL
Anbieter: Merck
Nitrate standards for ISE calibration, Orion™
Anbieter: Thermo Orion
Supelco® Organic Reference Standard, α-Arbutin
Anbieter: Merck
BREVETOXIN 3 SOLUTION 1 * 0,5 mL
Anbieter: Merck
Pharmaceutical secondary standard, Guaiacol, (CRM), Supelco®
Anbieter: Merck
IMAZAPYR CERTIFIED 1 * 0,1 g
Anbieter: EHRENSTORFER
2-Butanol, Reference Standard
Anbieter: USP