Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
GAMMA-BUTYROLACTONE (GBL) 1.0 MG/ML IN ACETONITRILE 1 * 1 mL
Catalog # PROOAMP1275.51-11
Supplier:
LGC Standards PROMOCHEM
Expand image
GAMMA-BUTYROLACTONE (GBL) 1.0 MG/ML IN ACETONITRILE 1 * 1 mL
Catalog # PROOAMP1275.51-11
Supplier:
LGC Standards PROMOCHEM
Frequently Bought Together
VWR® AVS TITRINORM Pufferlösungen, 20 °C
Supplier: VWR Chemicals
Pufferlösung, Ready to Use
Supplier: WTW
Pufferlösungen, Certipur®, farbcodiert, Supelco®
Supplier: Merck
Nitrat-Standardlösung, 1.000 mg/l Nitrat in Wasser (aus NaNO<sub>3</sub>) Certipur®, Supelco®
Supplier: Merck
Pufferlösungen, gebrauchsfertig, Referenztemperatur 20 °C, Certipur®, Supelco®
Supplier: Merck
Kaliumchlorid 3 mol/l (3 N) in wässriger Lösung, Supelco®
Supplier: Merck
pH Pufferlösungen, DuraCal
Supplier: HAMILTON BONADUZ
Leitfähigkeitsstandardlösungen
Supplier: VWR Chemicals
Leitfähigkeitsstandards bei 25 °C, Kaliumchloridlösungen
Supplier: VWR Chemicals
Pufferlösungen Certipur®, gebrauchsfertig, 1-Liter-Flaschen, Supelco®
Supplier: Merck
Pufferlösungen in Beuteln, Certipur® , Supelco®
Supplier: Merck