SFr. 0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
NAPHYRONE HYDROCHLORIDE (1-NAPHTHALEN-2-YL-2-PYRROLIDIN-1-YLPENTAN-1-ONE HYDROCHLORIDE) 1.0 MG/ML IN METHANOL (AS FREE BASE) 1 * 1 mL
PROOAMP1275.06-01
:
LGC Standards PROMOCHEM
NAPHYRONE HYDROCHLORIDE (1-NAPHTHALEN-2-YL-2-PYRROLIDIN-1-YLPENTAN-1-ONE HYDROCHLORIDE) 1.0 MG/ML IN METHANOL (AS FREE BASE) 1 * 1 mL
PROOAMP1275.06-01
:
LGC Standards PROMOCHEM
Frequently Bought Together
Sealing film for manual application, Parafilm® M
: HEATHROW
Aceton ≥99%, TECHNICAL
: VWR Chemicals
VWR® Gesichtstücher
: VWR Collection
VWR®, Nitrilhandschuhe
: VWR Collection
pH-Indikatorstäbchen, MQuant®, Supelco®
: Merck
Acetonitril ≥99,9%, HiPerSolv CHROMANORM® für die LC-MS, geeignet für UPLC/UHPLC-Instrumente
: VWR Chemicals
Acetonitril ≥99,9%, LiChrosolv® Reag. Ph. Eur., Gradienten-Qualität für die Flüssigchromatographie, Supelco®
: Merck
Wasser, HiPerSolv CHROMANORM® für die HPLC
: VWR Chemicals
Methanol ≥99,8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., Gradienten-Qualität für die HPLC
: VWR Chemicals
VWR®, Serologische Pipetten, Standard Serie
: VWR Collection
Nitrilhandschuhe, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® NITRILE LIGHT, Nitrilhandschuhe
: VWR Collection
VWR®, Transferpipetten, High Performance
: VWR Collection
Acetonitril, wasserfrei (max. 0,003% H₂O) ≥99,95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, Supergradienten-Qualität geeignet für UPLC/UHPLC-Instrumente
: VWR Chemicals