ContactUsLinkComponent
Clog Boxer 21P CH with mobile and adjustable strap, unisex, latex-free, autoclavable at 134C, with steel toecap 200J, antistatic sole, white, double size 41/42. EN ISO 20345:2011 A E SRC 1 * 1 PAAR
Catalog # BSNCBX21PCHCMBN42
Supplier:  Basan
Clog Boxer 21P CH with mobile and adjustable strap, unisex, latex-free, autoclavable at 134C, with steel toecap 200J, antistatic sole, white, double size 41/42. EN ISO 20345:2011 A E SRC 1 * 1 PAAR
Catalog # BSNCBX21PCHCMBN42
Supplier:  Basan
Frequently Bought Together