Mobile storage cabinets
Mobile storage cabinet, with 4 drawer of 150 mm height, lockable, 4 drawers, 900×516×740 mm
Catalog # KOTT315-00142
Supplier: KOETTERMANN
Jump to:
Product Details & Documents
Epoxy powder coated steel cabinets
- Optional locks
- Fitted with castors
- Removable shelves
Specifications
- Bezeichnung:Mobile storage cabinet, with 4 drawer of 150 mm height, lockable
- Seq:36
- B×T×H:900×516×740 mm
- Türen:4 drawers
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



