Mobile storage cabinets
Mobile storage cabinet, with 1 drawer of 150 mm height, 2 door, 900×516×740 mm
KOTT315-00138
Epoxy powder coated steel cabinets
- Optional locks
- Fitted with castors
- Removable shelves
- Bezeichnung:Mobile storage cabinet, with 1 drawer of 150 mm height
- Seq:32
- B×T×H:900×516×740 mm
- Türen:2 door
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



