Mobile storage cabinets
Mobile storage cabinet, with 1 drawer of 150 mm height and 1 drawer of 300 mm height, lockable, pencil tray, file drawer, 2 drawers, 450×516×590 mm
Katalognr. KOTT315-00109
Anbieter: KOETTERMANN
Direkt zu
Product Details & Documents
Epoxy powder coated steel cabinets
- Optional locks
- Fitted with castors
- Removable shelves
Technische Daten
- Bezeichnung:Mobile storage cabinet, with 1 drawer of 150 mm height and 1 drawer of 300 mm height, lockable, pencil tray, file drawer
- Seq:03
- B×T×H:450×516×590 mm
- Türen:2 drawers