Methylhexanoat 99%
Glasflasche, 250 g
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
Synonyme: Caproic acid methyl ester, Methyl caproate, Hexanoic acid methyl ester
- CAS Nummer:106-70-7
- Formel:C₇H₁₄O₂
- MDL-Nummer:MFCD00009510
- Molekulargewicht:130.19 g/mol
- Siedepunkt:151 °C (760 mmHg)
- Schmelzender Platingehalt:–71 °C
- Dichte:0,884 g/cm³
- Blitz Pt:42 °C
- Lagertemperatur:Raumtemperatur
- UN:3272
- ADR:3,III
Technische Daten
Ergebnisse des Spezifikationstests
- Appearance:Colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4040 to 1.4060 (20°C, 589 nm)
- GC:≥98.5 %
Technische Daten
- Packung:Glasflasche
- VE:250 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



