Methylhexanoat 99%
Glasflasche, 50 g
: Caproic acid methyl ester, Methyl caproate, Hexanoic acid methyl ester
- :106-70-7
- :C₇H₁₄O₂
- :MFCD00009510
- :130.19 g/mol
- :151 °C (760 mmHg)
- :–71 °C
- :0,884 g/cm³
- :42 °C
- :Raumtemperatur
- :3272
- :3,III
- Appearance:Colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4040 to 1.4060 (20°C, 589 nm)
- GC:≥98.5 %
- Packung:Glasflasche
- VE:50 g



