Methylhexanoat 99%
Glasflasche, 1 kg
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Caproic acid methyl ester, Methyl caproate, Hexanoic acid methyl ester
- CAS Number:106-70-7
- Formula:C₇H₁₄O₂
- MDL Number:MFCD00009510
- Molecular Weight:130.19 g/mol
- Boiling Pt:151 °C (760 mmHg)
- Melting Pt:–71 °C
- Density:0,884 g/cm³
- Flash Pt:42 °C
- Storage Temperature:Raumtemperatur
- UN:3272
- ADR:3,III
Documents
ACRO44924
SDS
Specifications
Specification Test Results
- Appearance:Colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4040 to 1.4060 (20°C, 589 nm)
- GC:≥98.5 %
Specifications
- Packung:Glasflasche
- VE:1 kg



