Methyllinoleat 99%
Glasflasche, 100 g
: cis,cis-9,12-Octadecadienoic acid methyl ester, 9,12-Octadecanoic acid methyl ester, 9Z,12Z-Octadecadienoic acid methyl ester, Linoleic acid methyl ester, (9Z,12Z)-Methyl octadeca-9,12-dienoate, Methyl cis,cis-9,12-octadecadienoate, 9Z,12Z-Oktadekadiensäuremethylester, 9,12 Methyl octadecaotenoate methyl linoleate, cis,cis-9,12-Oktadekadiensäuremethylester
- :112-63-0
- :C₁₉H₃₄O₂
- :MFCD00009534
- :294.48 g/mol
- :212 °C (16 mmHg)
- :–35 °C
- :0,89 g/cm³
- :>200 °C
- :Kühlschrank
- :203-993-0
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4610 to 1.4630 (20°C, 589 nm)
- GC:≥98.5 %
- Specific optical rotation:-85° to -95° (20°C, 589 nm) (c=1, CHCl3)
- Packung:Glasflasche
- VE:100 g



