Schwefelsäure 95% ACS
Kst.-Flasche, 500 mL
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
- CAS Nummer:7664-93-9
- Formel:H₂SO₄
- MDL-Nummer:MFCD00064589
- Molekulargewicht:98.08 g/mol
- Merck-Index:14,08974
- Siedepunkt:∼335 °C (1013 hPa)
- Schmelzender Platingehalt:10 °C
- Dichte:1,83 g/cm³
- Lagertemperatur:Raumtemperatur
- EINECS:231-639-5
- ADR:8,II
- :Density: 1.84 g/cm³ (20 °C)
- UN:1830
Documents
Technische Daten
Ergebnisse des Spezifikationstests
- Appearance:Free from suspended or insoluble matter
- Color scale:≤10 APHA
- Residue after ignition:≤0.0005 %
- Titration with NaOH:95.0 to 98.0 %
- Chloride (Cl):≤0.2 ppm
- Nitrate (NO3):≤0.5 ppm
- Ammonium (NH4):≤2 ppm
- Arsenic (As):≤0.01 ppm
- Iron (Fe):≤0.2 ppm
- Mercury (Hg):≤5 ppb
- Heavy metals (as Pb):≤1 ppm
- KMnO4-reducing substance:≤2 ppm (as SO2)
Technische Daten
- Packung:Kst.-Flasche
- VE:500 mL
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



