Paraldehyd 99.5%
Glasflasche, 2,5 L
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
- CAS Nummer:123-63-7
- Formel:C₆H₁₂O₃
- MDL-Nummer:MFCD00036208
- Molekulargewicht:132.16 g/mol
- Schmelzender Platingehalt:12 °C
- Dichte:0,99 g/cm³
- Blitz Pt:24 °C
- Lagertemperatur:Raumtemperatur
- EINECS:204-639-8
- UN:1264
- ADR:3,III
Documents
ACRO27508
FDS
Technische Daten
Ergebnisse des Spezifikationstests
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4035 to 1.4055 (20°C, 589 nm)
- Water:≤0.1 % (K.F.)
- GC:≥99.4 %
- Free acid (CH3COOH):≤0.3 % (titr.)
Technische Daten
- Packung:Glasflasche
- VE:2,5 L
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



