Rosolsäure (Aurin)
Glasflasche, 500 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: 4-(Bis(4-hydroxyphenyl)methylene)cyclohexa-2,5-dien-1-one
- CAS Number:603-45-2
- Formula:C₁₉H₁₄O₃
- MDL Number:MFCD00001624
- Molecular Weight:290.32 g/mol
- Storage Temperature:Raumtemperatur
- EINECS:210-041-8
Documents
ACRO22983
SDS
Specifications
Specification Test Results
- Appearance:Red-brown to brown Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤10 % (110°C, 1 h)
- UV Lambda max:480 to 486 nm (in ethanol + 2ml 1N HCl)
- Visual transition interval:from pH 6.2 (yellow) to pH 8.2 (red-pink)
Specifications
- Packung:Glasflasche
- VE:500 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



