Methyllaurat 98+%
Glasflasche, 250 g
: Dodecylic acid methyl ester, Methyl dodecanoate, Lauric acid methyl ester, Dodecanoic acid methyl ester
- :111-82-0
- :C₁₃H₂₆O₂
- :MFCD00008966
- :214.35 g/mol
- :262 °C (766 mmHg)
- :4…5 °C
- :0,87 g/cm³
- :>110 °C
- :Raumtemperatur
- :203-911-3
- :3082
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4290 to 1.4340 (20°C, 589 nm)
- GC:≥98.0 %
- Packung:Glasflasche
- VE:250 g
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



