Methyllaurat 98+%
Glasflasche, 50 g
: Dodecylic acid methyl ester, Methyl dodecanoate, Lauric acid methyl ester, Dodecanoic acid methyl ester
- :111-82-0
- :C₁₃H₂₆O₂
- :MFCD00008966
- :214.35 g/mol
- :262 °C (766 mmHg)
- :4…5 °C
- :0,87 g/cm³
- :>110 °C
- :Raumtemperatur
- :203-911-3
- :3082
Documents
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4290 to 1.4340 (20°C, 589 nm)
- GC:≥98.0 %
- Packung:Glasflasche
- VE:50 g



