Methyllaurat 98+%
Glasflasche, 10 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Dodecylic acid methyl ester, Methyl dodecanoate, Lauric acid methyl ester, Dodecanoic acid methyl ester
- CAS Number:111-82-0
- Formula:C₁₃H₂₆O₂
- MDL Number:MFCD00008966
- Molecular Weight:214.35 g/mol
- Boiling Pt:262 °C (766 mmHg)
- Melting Pt:4…5 °C
- Density:0,87 g/cm³
- Flash Pt:>110 °C
- Storage Temperature:Raumtemperatur
- EINECS:203-911-3
- UN:3082
Documents
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4290 to 1.4340 (20°C, 589 nm)
- GC:≥98.0 %
Specifications
- Packung:Glasflasche
- VE:10 g



