Kupfer(II)oxid 99+%
Kst.-Flasche, 2,5 kg
: Cupric oxide, Copper oxide, Copper monoxide granulated
- :1317-38-0
- :CuO
- :MFCD00010979
- :79.55 g/mol
- :13,02674
- :2000 °C (1013 hPa)
- :1326 °C
- :4,9 g/cm³ (20 °C)
- :Raumtemperatur
- :215-269-1
- :3077
- :9,III
- Appearance:Black Powder
- Titration Complexometric:≥99.0 %
- Chloride (Cl):≤50 ppm
- Carbon (C):≤500 ppm
- Iron (Fe):≤500 ppm
- Total sulfur (as SO4):≤0.02 %
- Free alkali:Passes test
- Insoluble matter:≤0.02 % (in dilute HCl)
- Nitrogen compounds (as N):≤0.002 %
- Packung:Kst.-Flasche
- VE:2,5 kg



