L(+)-Cystein 98+%
Kst.-Flasche, 500 g
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
Synonyme: H-Cys-OH, L-(+)-Cysteine, (R)-2-Amino-3-mercaptopropanoic acid
- CAS Nummer:52-90-4
- Formel:HSCH₂CH(NH₂)CO₂H
- MDL-Nummer:MFCD00064306
- Molekulargewicht:121.16 g/mol
- Merck-Index:13,02810
- Siedepunkt:254 °C (1013 hPa)
- Schmelzender Platingehalt:220 °C (Zers.)
- Schmelzender Platingehalt:220 °C (Zersetzung)
- Dichte:1,523 g/cm³ (20 °C)
- Lagertemperatur:Kühlschrank
- EINECS:200-158-2
Documents
Technische Daten
Ergebnisse des Spezifikationstests
- Appearance:White to almost white Powder or crystals
- Infrared spectrum:Conforms
- Loss on drying:≤0.5 % (105°C, 3 hrs)
- Solubility:(2.5 % in water) Clear colorless
- Residue after ignition:≤0.1 %
- pH:4.5 to 5.5 (2.5 % in water)
- Specific optical rotation:+7.5° to +9.5° (20°C, 589 nm) (c=8, 5N HCl)
- Titration with HClO4:98.0 to 102.0 %
- Chloride (Cl):≤0.05 %
- Sulfate (SO4):≤0.03 %
- Ammonium (NH4):≤0.02 %
- Iron (Fe):≤10 ppm
- Heavy metals (as Pb):≤10 ppm
Technische Daten
- Packung:Kst.-Flasche
- VE:500 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



